Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp4a12b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172306.2 | NP_758510.2 | 4, 5 | 571 | ||
GenBank mRNA | AK098123.1 | - | 4, 5 | 552 | ||
BC060945.1 | - | 4, 5 | 552 | 30304458 |
Gene Symbol: Cyp4a12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177406.3 | NP_803125.2 | 4, 5 | 594 | ||
GenBank mRNA | AK144260.1 | - | 4, 5 | 524 | ||
BC014721.1 | - | 4, 5 | 540 | 4240359 | ||
BC025936.1 | - | 4, 5 | 534 | 5097698 | ||
BC026582.1 | - | 4, 5 | 540 | 4971335 | ||
BC031141.1 | - | 4, 5 | 515 | 4972684 | ||
BC033924.1 | - | 4, 5 | 524 | 5101821 | ||
CB955255.1 | - | 4, 5 | 596 |