Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HCLS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001292041.1 | NP_001278970.1 | 13 | 1378 | ||
NM_005335.5 | NP_005326.2 | 14 | 1489 | |||
GenBank mRNA | AK124015.1 | - | 9 | 2175 | ||
AK225633.1 | - | 13 | 1236 | |||
AK298696.1 | - | 13 | 1328 | |||
AK312750.1 | - | 14 | 1452 | |||
BC016758.2 | - | 14 | 1409 | 2966303 | ||
BT006824.1 | - | 13 | 1360 | |||
CR456794.1 | - | 13 | 1360 | |||
X16663.1 | - | 14 | 1402 |