Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dbi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037999.2 | NP_001033088.1 | 4 | 536 | ||
NM_007830.4 | NP_031856.1 | 4 | 530 | |||
GenBank mRNA | AK008540.1 | - | 4 | 451 | ||
AK008576.1 | - | 4 | 451 | |||
AK018720.1 | - | 4 | 452 | |||
AK020692.1 | - | 4 | 500 | |||
AK027906.1 | - | 4 | 452 | |||
AK078428.1 | - | 4 | 514 | |||
AK145278.1 | - | 4 | 452 | |||
AK148753.1 | - | 4 | 453 | |||
AV001592.1 | - | 4 | 424 | |||
BC028874.1 | - | 4 | 430 | 4194069 | ||
EF437365.1 | - | 4 | 437 |