Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007832.4 | NP_031858.1 | 3, 4 | 652 | ||
GenBank mRNA | AK004592.1 | - | 3, 4 | 477 | ||
AK010700.1 | - | 3, 4 | 442 | |||
AK048347.1 | - | 3, 4 | 489 | |||
AK088195.1 | - | 3, 4 | 489 | |||
AK145353.1 | - | 4, 5 | 598 | |||
AK145390.1 | - | 4, 5 | 598 | |||
AK151851.1 | - | 3, 4 | 486 | |||
BC052004.1 | - | 3, 4 | 576 | 6401517 | ||
BC060062.1 | - | 3, 4 | 638 | 5703508 | ||
BU052737.1 | - | 3, 4 | 654 | |||
X77731.1 | - | 3, 4 | 421 |