Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dct | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010024.3 | NP_034154.2 | 1 | 694 | ||
GenBank mRNA | AK033040.1 | - | 1 | 693 | ||
AK178101.1 | - | 1 | 102 | |||
AK187215.1 | - | 1 | 102 | |||
AK190334.1 | - | 1 | 102 | |||
AK193217.1 | - | 1 | 102 | |||
AK198559.1 | - | 1 | 102 | |||
AK200366.1 | - | 1 | 102 | |||
AK209892.1 | - | 1 | 352 | |||
AK212164.1 | - | 1 | 272 | |||
AK214181.1 | - | 1 | 270 | |||
AK214448.1 | - | 1 | 253 | |||
AK219002.1 | - | 1 | 281 | |||
BC067064.1 | - | 1 | 528 | 30537997 | ||
BC082330.1 | - | 1 | 329 | 30539879 | ||
EU554632.1 | - | 1 | 226 | |||
X63349.1 | - | 1 | 630 |