Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dct | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010024.3 | NP_034154.2 | 6 | 1524 | ||
GenBank mRNA | AK033040.1 | - | 6 | 1523 | ||
AK177534.1 | - | 2 | 176 | |||
AK186214.1 | - | 2 | 174 | |||
AK190354.1 | - | 2 | 131 | |||
AK191069.1 | - | 2 | 148 | |||
AK191107.1 | - | 2 | 146 | |||
AK192618.1 | - | 2 | 174 | |||
AK192803.1 | - | 2 | 174 | |||
AK195046.1 | - | 2 | 174 | |||
AK195075.1 | - | 2 | 129 | |||
AK196076.1 | - | 2 | 126 | |||
AK196684.1 | - | 2 | 174 | |||
AK198785.1 | - | 2 | 132 | |||
AK198901.1 | - | 2 | 175 | |||
AK199676.1 | - | 2 | 174 | |||
AK199862.1 | - | 2 | 174 | |||
AK200015.1 | - | 2 | 139 | |||
AK201463.1 | - | 2 | 174 | |||
AK201739.1 | - | 2 | 174 | |||
AK202339.1 | - | 2 | 174 | |||
AK205161.1 | - | 2 | 122 | |||
AK205196.1 | - | 2 | 119 | |||
AK207218.1 | - | 2 | 174 | |||
AK208819.1 | - | 2 | 174 | |||
AK210502.1 | - | 2 | 174 | |||
AK214235.1 | - | 2 | 174 | |||
AK216743.1 | - | 2 | 148 | |||
BC067064.1 | - | 6 | 1358 | 30537997 | ||
BC082330.1 | - | 6 | 1159 | 30539879 | ||
EU554632.1 | - | 6 | 1056 | |||
X63349.1 | - | 6 | 1460 |