Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110826.1 | NP_001104296.1 | 12 | 1619 | ||
NM_007841.4 | NP_031867.1 | 12 | 1562 | |||
NM_181324.3 | NP_851841.2 | 12 | 1587 | |||
GenBank mRNA | AK054144.1 | - | 12 | 1581 | ||
AK148483.1 | - | 12 | 1584 | |||
BC021452.1 | - | 11 | 1479 | 5043599 |