Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc29a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007854.3 | NP_031880.2 | 6, 7 | 892 | ||
GenBank mRNA | AF183397.1 | - | 6, 7 | 802 | ||
AK010950.1 | - | 6, 7 | 829 | |||
AK083484.1 | - | 6, 7 | 789 | |||
AK154310.1 | - | 6, 7 | 827 | |||
BC048958.1 | - | 6, 7 | 1038 | 6491511 | ||
X86682.1 | - | 4, 5 | 460 |