Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ackr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010045.2 | NP_034175.2 | 2 | 522 | ||
GenBank mRNA | AF016584.1 | - | 2 | 463 | ||
AF109159.1 | - | 2 | 522 | |||
AY289754.1 | - | 2 | 429 | |||
AY291295.1 | - | 2 | 429 | |||
BC005583.1 | - | 2 | 521 | 3968984 |