Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dio2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010050.2 | NP_034180.1 | 1 | 359 | ||
GenBank mRNA | AF093137.1 | - | 1 | 217 | ||
AF096875.1 | - | 1 | 359 | |||
AF177196.1 | - | 1 | 229 | |||
AF177197.1 | - | 1 | 229 | |||
AK032038.1 | - | 1 | 69 | |||
AK045607.1 | - | 1 | 113 | |||
BC125383.1 | - | 1 | 254 | 40129898 | ||
BC125385.1 | - | 1 | 254 | 40129900 |