Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dld | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007861.5 | NP_031887.2 | 7, 8 | 721 | ||
GenBank mRNA | AK117104.1 | - | 7, 8 | 665 | ||
AK135807.1 | - | 7, 8 | 664 | |||
AK136193.1 | - | 7, 8 | 665 | |||
AK153399.1 | - | 7, 8 | 665 | |||
AK167885.1 | - | 7, 8 | 665 | |||
AK168157.1 | - | 7, 8 | 686 | |||
AK168875.1 | - | 7, 8 | 606 | |||
AK193031.1 | - | 1, 2 | 121 | |||
BC003368.1 | - | 7, 8 | 648 | 3589344 | ||
U73445.1 | - | 7, 8 | 698 |