Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007867.4 | NP_031893.3 | 3 | 1470 | ||
GenBank mRNA | AK145123.1 | - | 3 | 1933 | ||
U73329.1 | - | 3 | 1150 |