Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnpep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110831.1 | NP_001104301.1 | 12 | 1392 | ||
NM_016878.4 | NP_058574.3 | 12 | 1216 | |||
GenBank mRNA | AK038646.1 | - | 1392 | |||
AK052001.1 | - | 1194 | ||||
AK160084.1 | - | 12 | 1153 | |||
AK166276.1 | - | 12 | 1166 | |||
BC092232.1 | - | 12 | 1144 | 30551654 |