Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnpep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110831.1 | NP_001104301.1 | 13 | 1420 | ||
NM_016878.4 | NP_058574.3 | 13 | 1244 | |||
GenBank mRNA | AF005051.1 | - | 12 | 1354 | ||
AK038646.1 | - | 1420 | ||||
AK052001.1 | - | 1222 | ||||
AK160084.1 | - | 13 | 1181 | |||
AK166276.1 | - | 13 | 1194 | |||
AK182346.1 | - | 4 | 238 | |||
AK188773.1 | - | 2 | 34 | |||
BC092232.1 | - | 13 | 1172 | 30551654 |