Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Doc2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007873.2 | NP_031899.2 | 4 | 757 | ||
GenBank mRNA | BC067030.1 | - | 4 | 757 | 30534150 | |
D85037.1 | - | 4 | 749 |