Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Drd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010077.2 | NP_034207.2 | 5 | 717 | ||
GenBank mRNA | BC105663.1 | - | 5 | 717 | 40046665 | |
BC105664.1 | - | 5 | 717 | 40046668 | ||
BC105665.1 | - | 5 | 717 | 40046660 | ||
BC105666.1 | - | 5 | 717 | 40046662 | ||
X55674.1 | - | 5 | 752 |
Set Membership: |
G-protein coupled receptor |