Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Drp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010078.3 | NP_034208.2 | 23 | 3016 | ||
GenBank mRNA | AK081426.1 | - | 23 | 2960 | ||
AK158102.1 | - | 22 | 2818 | |||
BC060605.1 | - | 21 | 3595 | |||
BC125345.1 | - | 23 | 2876 | 40129860 | ||
BC125347.1 | - | 23 | 2876 | 40129862 |