Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dsc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013505.3 | NP_038533.1 | 13 | 2200 | ||
GenBank mRNA | AK028604.1 | - | 14 | 2240 | ||
AK028636.1 | - | 13 | 2201 | |||
AK030333.1 | - | 13 | 2201 | |||
AK048547.1 | - | 13 | 2201 | |||
AK172168.1 | - | 13 | 2035 | |||
BC004663.1 | - | 7 | 1087 | |||
BC057867.1 | - | 13 | 1980 | 5327748 | ||
L33779.1 | - | 13 | 1999 |