Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000186.3 | NP_000177.2 | 4 | 649 | ||
NM_001014975.2 | NP_001014975.1 | 4 | 649 | |||
XM_017001108.1 | XP_016856597.1 | 4 | 531 | |||
GenBank mRNA | AK225649.1 | - | 4 | 489 | ||
AK291395.1 | - | 4 | 488 | |||
BC012610.1 | - | 4 | 475 | |||
BC037285.2 | - | 4 | 474 | |||
BC110643.1 | - | 4 | 465 | |||
BC142699.1 | - | 4 | 461 | 40148771 | ||
X04697.1 | - | 3 | 253 | |||
X07523.1 | - | 4 | 482 | |||
Y00716.1 | - | 4 | 482 |