Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000186.3 | NP_000177.2 | 2 | 461 | ||
NM_001014975.2 | NP_001014975.1 | 2 | 461 | |||
XM_017001108.1 | XP_016856597.1 | 2 | 343 | |||
GenBank mRNA | AK225649.1 | - | 2 | 301 | ||
AK291395.1 | - | 2 | 300 | |||
BC012610.1 | - | 2 | 287 | |||
BC037285.2 | - | 2 | 286 | |||
BC110643.1 | - | 2 | 277 | |||
BC142699.1 | - | 2 | 273 | 40148771 | ||
BM842566.1 | - | 2 | 461 | |||
X04697.1 | - | 1 | 65 | |||
X07523.1 | - | 2 | 294 | |||
Y00716.1 | - | 2 | 294 |