Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dst | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276764.1 | NP_001263693.1 | 47 | 12310 | ||
NM_133833.3 | NP_598594.2 | 39 | 5687 | |||
NM_134448.3 | NP_604443.2 | 44 | 11729 | |||
GenBank mRNA | AF396878.1 | - | 39 | 5699 | ||
AF396879.1 | - | 44 | 11726 |