Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dst | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276764.1 | NP_001263693.1 | 51 | 12997 | ||
NM_133833.3 | NP_598594.2 | 43 | 6374 | |||
NM_134448.3 | NP_604443.2 | 48 | 12416 | |||
GenBank mRNA | AF396878.1 | - | 43 | 6386 | ||
AF396879.1 | - | 48 | 12413 | |||
BU919808.1 | - | 4 | 431 |