Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000186.3 | NP_000177.2 | 8 | 1245 | ||
NM_001014975.2 | NP_001014975.1 | 8 | 1245 | |||
XM_017001108.1 | XP_016856597.1 | 8 | 1127 | |||
GenBank mRNA | AK226113.1 | - | 3 | 3031 | ||
BC012610.1 | - | 7 | 879 | |||
BC037285.2 | - | 8 | 1070 | |||
BC110643.1 | - | 8 | 1061 | |||
BC142699.1 | - | 8 | 1057 | 40148771 | ||
M12383.1 | - | 3 | 330 | |||
X04697.1 | - | 7 | 849 | |||
X07523.1 | - | 8 | 1078 | |||
Y00716.1 | - | 8 | 1078 |