Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048175.2 | NP_001041640.1 | 12, 13 | 1447 | ||
NM_010082.2 | NP_034212.1 | 12, 13 | 1447 | |||
NM_183366.3 | NP_899222.1 | 12, 13 | 1447 | |||
GenBank mRNA | AF153350.1 | - | 12, 13 | 1354 | ||
AF163290.1 | - | 12, 13 | 1447 | |||
AF163291.1 | - | 12, 13 | 1447 | |||
AF163292.1 | - | 12, 13 | 1447 | |||
AF163293.1 | - | 12, 13 | 1447 | |||
AK136715.1 | - | 3, 4 | 351 | |||
BC058782.1 | - | 12, 13 | 1322 | 6310763 | ||
U06146.1 | - | 14 |