Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010084.2 | NP_034214.2 | 17 | 1889 | ||
GenBank mRNA | AF167405.1 | - | 17 | 1887 | ||
AK076633.1 | - | 17 | 1893 | |||
BC051136.1 | - | 17 | 1866 | 6708158 | ||
BC144841.1 | - | 17 | 1862 | |||
BC145751.1 | - | 17 | 1862 | 40131054 |