Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dtnb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162465.1 | NP_001155937.1 | 4 | 423 | ||
NM_007886.2 | NP_031912.2 | 4 | 358 | |||
GenBank mRNA | AJ003007.1 | - | 4 | 291 | ||
AK043436.1 | - | 4 | 381 | |||
AK083752.1 | - | 4 | 379 | |||
AK157158.1 | - | 4 | 380 | |||
BC030924.1 | - | 4 | 358 | 4910166 | ||
Y15742.1 | - | 4 | 255 |