Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dtnb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162465.1 | NP_001155937.1 | 3 | 357 | ||
NM_007886.2 | NP_031912.2 | 3 | 292 | |||
GenBank mRNA | AJ003007.1 | - | 3 | 225 | ||
AK043436.1 | - | 3 | 315 | |||
AK083752.1 | - | 3 | 313 | |||
AK085675.1 | - | 3 | 282 | |||
AK157158.1 | - | 3 | 314 | |||
BC030924.1 | - | 3 | 292 | 4910166 | ||
Y15742.1 | - | 3 | 189 |