Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp17la | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007887.2 | NP_031913.1 | 3 | 1271 | ||
GenBank mRNA | BC140988.1 | - | 3 | 1300 | 9055600 | |
U41636.1 | - | 3 | 1283 |