Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dvl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007889.2 | NP_031915.2 | 4 | 516 | ||
GenBank mRNA | AK052151.1 | - | 4 | 517 | ||
AK199514.1 | - | 2 | 125 | |||
AK220155.1 | - | 2 | 3915 | |||
BC033414.1 | - | 3 | 1478 | |||
U41285.1 | - | 4 | 598 |