Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dyrk1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113389.1 | NP_001106860.1 | 10 | 1493 | ||
NM_007890.2 | NP_031916.1 | 10 | 1752 | |||
GenBank mRNA | AK052390.1 | - | 4 | 628 | ||
BC034550.1 | - | 5 | 838 | |||
BC048837.1 | - | 1 | 47 | |||
BC129888.1 | - | 9 | 1355 | 40111469 | ||
BC129889.1 | - | 9 | 1355 | 40111470 | ||
BC138716.1 | - | 9 | 1382 | 8861738 | ||
BC145405.1 | - | 9 | 1355 | 9053963 | ||
U58497.1 | - | 9 | 1382 |