Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapre1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007896.3 | NP_031922.1 | 3 | 360 | ||
GenBank mRNA | AK013626.1 | - | 3 | 299 | ||
AK146206.1 | - | 3 | 278 | |||
AK154245.1 | - | 3 | 304 | |||
BC043333.1 | - | 3 | 324 | |||
BC052405.1 | - | 2 | 235 | |||
BC052728.1 | - | 271 | ||||
BC064444.1 | - | 271 | 6815724 | |||
U51196.1 | - | 3 | 290 |