Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ebp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK012266.1 | - | 3, 4 | 493 | ||
AK027977.1 | - | 3, 4 | 465 | |||
BC004620.1 | - | 3, 4 | 471 | 3585323 | ||
BC004703.1 | - | 3, 4 | 1103 | 3499265 | ||
DV661455.1 | - | 2, 3 | 67 | |||
X97755.1 | - | 3, 4 | 469 |