Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sparcl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010097.4 | NP_034227.3 | 6 | 1987 | ||
GenBank mRNA | AK134608.1 | - | 6 | 1628 | ||
AK158725.1 | - | 6 | 1494 | |||
BC003759.1 | - | 6 | 1632 | 3486162 | ||
U64827.1 | - | 6 | 1585 | |||
U77330.1 | - | 6 | 1652 |