Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HGF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000601.5 | NP_000592.3 | 5 | 754 | ||
NM_001010931.2 | NP_001010931.1 | 5 | 754 | |||
NM_001010932.2 | NP_001010932.1 | 5 | 739 | |||
NM_001010933.2 | NP_001010933.1 | 5 | 739 | |||
NM_001010934.2 | NP_001010934.1 | 5 | 754 | |||
XM_006715956.2 | XP_006716019.1 | 6 | 717 | |||
XM_011516115.2 | XP_011514417.1 | 6 | 702 | |||
XM_017012097.1 | XP_016867586.1 | 6 | 717 | |||
XM_017012098.1 | XP_016867587.1 | 6 | 702 | |||
GenBank mRNA | AB208900.1 | - | 5 | 661 | ||
AK291674.1 | - | 5 | 689 | |||
AK313694.1 | - | 5 | 589 | |||
BC022308.1 | - | 5 | 588 | |||
BC063485.1 | - | 5 | 612 | |||
BC105797.1 | - | 5 | 694 | |||
BC130284.1 | - | 5 | 602 | 40146315 | ||
BC130286.1 | - | 5 | 602 | 40146317 | ||
L02931.1 | - | 5 | 587 | |||
M29145.1 | - | 5 | 628 | |||
M55379.1 | - | 5 | 512 | |||
M60718.1 | - | 5 | 599 | |||
M73239.1 | - | 5 | 592 | |||
M73240.1 | - | 5 | 577 | |||
M77227.1 | - | 5 | 609 | |||
U46010.1 | - | 5 | 581 | |||
X16323.1 | - | 5 | 661 | |||
X57574.1 | - | 5 | 604 |