Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: S1pr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010101.4 | NP_034231.1 | 2 | 1489 | ||
GenBank mRNA | AB028143.1 | - | 2 | 1357 | ||
AK028043.1 | - | 2 | 1346 | |||
AK029852.1 | - | 2 | 1345 | |||
AK030134.1 | - | 2 | 1344 | |||
AK047268.1 | - | 2 | 1337 | |||
AK078443.1 | - | 2 | 1343 | |||
AK081919.1 | - | 2 | 1334 | |||
AK083405.1 | - | 2 | 1323 | |||
AK084944.1 | - | 2 | 1344 | |||
AK085180.1 | - | 2 | 1344 | |||
AK087076.1 | - | 2 | 1345 | |||
AK087139.1 | - | 2 | 1343 | |||
BC064007.1 | - | 2 | 1329 | 30244496 |
Set Membership: |
G-protein coupled receptor |