Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162425.1 | NP_001155897.1 | 3 | 468 | ||
NM_010107.4 | NP_034237.3 | 3 | 504 | |||
GenBank mRNA | AK009144.1 | - | 3 | 501 | ||
AK079266.1 | - | 3 | 471 | |||
BC002046.1 | - | 3 | 469 | 3484518 | ||
CN696019.1 | - | 3 | 504 | |||
D38146.1 | - | 3 | 401 | |||
U26188.1 | - | 3 | 463 | |||
U90662.1 | - | 3 | 403 |