Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efnb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007911.5 | NP_031937.1 | 2 | 572 | ||
GenBank mRNA | AF025288.1 | - | 2 | 190 | ||
AK048305.1 | - | 2 | 516 | |||
BC052001.1 | - | 2 | 389 | 5695429 | ||
BC058617.1 | - | 2 | 505 | 5695418 |