Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Egf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010113.3 | NP_034243.2 | 12 | 2139 | ||
GenBank mRNA | AK085707.1 | - | 12 | 1706 | ||
AK088408.1 | - | 7 | 726 | |||
AK136439.1 | - | 7 | 732 | |||
AK142666.1 | - | 12 | 1835 | |||
AK157778.1 | - | 7 | 731 | |||
BC017681.1 | - | 13 | 2076 | 4221592 | ||
BC060741.1 | - | 12 | 2124 | 4164289 | ||
BC092277.1 | - | 12 | 2124 | 4240429 | ||
J00380.1 | - | 12 | 2134 | |||
V00741.1 | - | 12 | 2090 |