Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif4ebp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007918.3 | NP_031944.3 | 3 | 590 | ||
GenBank mRNA | AK013033.1 | - | 516 | |||
AK167836.1 | - | 3 | 591 | |||
AK169979.1 | - | 3 | 536 | |||
AK170031.1 | - | 3 | 540 | |||
BC002045.1 | - | 3 | 558 | 3592595 | ||
BE651550.1 | - | 3 | 481 | |||
U28656.1 | - | 3 | 558 |