Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif4g2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040131.2 | NP_001035221.1 | 18 | 2360 | ||
NM_013507.3 | NP_038535.2 | 19 | 2474 | |||
GenBank mRNA | AK144309.1 | - | 19 | 2448 | ||
AK150745.1 | - | 19 | 2451 | |||
AK151637.1 | - | 19 | 2451 | |||
BC039274.1 | - | 17 | 2038 | |||
BC040391.1 | - | 18 | 2163 | 4018962 | ||
BC043034.1 | - | 18 | 2154 | 5696921 | ||
BC056387.1 | - | 19 | 2438 | 6854755 | ||
BC057673.1 | - | 19 | 2212 | 4020759 | ||
BC064810.1 | - | 18 | 2207 | 6825556 | ||
BC092521.1 | - | 19 | 2429 | 30649919 | ||
BC127064.1 | - | 19 | 2474 | 40109978 | ||
CN700661.1 | - | 2 | 241 | |||
U76112.1 | - | 19 | 2448 |