Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif4g2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040131.2 | NP_001035221.1 | 11 | 1305 | ||
NM_013507.3 | NP_038535.2 | 11 | 1305 | |||
GenBank mRNA | AK141512.1 | - | 4 | 375 | ||
AK144309.1 | - | 11 | 1279 | |||
AK150745.1 | - | 11 | 1282 | |||
AK151637.1 | - | 11 | 1282 | |||
BC039274.1 | - | 10 | 983 | |||
BC040391.1 | - | 10 | 994 | 4018962 | ||
BC043034.1 | - | 10 | 985 | 5696921 | ||
BC056387.1 | - | 11 | 1269 | 6854755 | ||
BC057673.1 | - | 12 | 1157 | 4020759 | ||
BC064810.1 | - | 10 | 1038 | 6825556 | ||
BC092521.1 | - | 11 | 1260 | 30649919 | ||
BC127064.1 | - | 11 | 1305 | 40109978 | ||
BU610672.1 | - | 3 | 287 | |||
U63323.1 | - | 11 | 1257 | |||
U76112.1 | - | 11 | 1279 |