Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163131.1 | NP_001156603.1 | 8 | 1160 | ||
NM_007921.3 | NP_031947.1 | 8 | 1100 | |||
GenBank mRNA | AB221650.1 | - | 7 | 968 | ||
AF016294.1 | - | 8 | 1066 | |||
AK143135.1 | - | 8 | 1150 | |||
BC138692.1 | - | 8 | 1064 | 8861714 | ||
BC145380.1 | - | 8 | 1124 | 9053938 |