Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2K | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111112.1 | NP_001104582.1 | 3, 4 | 508 | ||
NM_001312647.1 | NP_001299576.1 | 3, 4 | 420 | |||
NM_001312648.1 | NP_001299577.1 | 2, 3 | 414 | |||
NM_005339.4 | NP_005330.1 | 3, 4 | 508 | |||
GenBank mRNA | AB022435.1 | - | 3, 4 | 310 | ||
AK093533.1 | - | 3, 4 | 329 | |||
AK307130.1 | - | 2, 3 | 319 | |||
AK315524.1 | - | 3, 4 | 434 | |||
BC022804.1 | - | 3, 4 | 384 | 4893778 | ||
BC050600.1 | - | 3, 4 | 362 | 6502901 | ||
CX786829.1 | - | 3, 4 | 347 | |||
DA526283.1 | - | 3, 4 | 508 | |||
U58522.1 | - | 3, 4 | 227 |