Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mark2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080388.2 | NP_001073857.1 | 13 | 1922 | ||
NM_001080389.2 | NP_001073858.1 | 13 | 1922 | |||
NM_001080390.2 | NP_001073859.1 | 13 | 1639 | |||
NM_007928.3 | NP_031954.2 | 13 | 1922 | |||
GenBank mRNA | AK045329.1 | - | 13 | 1496 | ||
AK154864.1 | - | 13 | 1639 | |||
AK162704.1 | - | 2 | 132 | |||
AK172444.1 | - | 13 | 1882 | |||
AK172664.1 | - | 13 | 1565 | |||
AK177415.1 | - | 2 | 152 | |||
AK184786.1 | - | 2 | 152 | |||
AK211690.1 | - | 4 | 370 | |||
AK220191.1 | - | 14 | 1488 | |||
BC058556.1 | - | 13 | 1859 | 5289799 | ||
CX210531.1 | - | 3 | 285 | |||
X70764.1 | - | 13 | 1370 |