Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: En2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010134.3 | NP_034264.1 | 2 | 1501 | ||
GenBank mRNA | AK157766.1 | - | 2 | 1503 | ||
L12705.1 | - | 2 | 717 |