Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Enah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083120.2 | NP_001076589.1 | 8, 9 | 2460 | ||
NM_001083121.2 | NP_001076590.1 | 8, 9 | 1728 | |||
NM_008680.3 | NP_032706.2 | 9, 10 | 2472 | |||
NM_010135.3 | NP_034265.2 | 9, 10 | 2517 | |||
GenBank mRNA | BC062927.1 | - | 8, 9 | 1347 | 6851217 | |
U72520.1 | - | 8, 9 | 1421 | |||
U72521.1 | - | 8, 9 | 2147 | |||
U72522.1 | - | 9, 10 | 2159 | |||
U72523.1 | - | 9, 10 | 2204 |