Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Enc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007930.4 | NP_031956.3 | 2 | 769 | ||
GenBank mRNA | AK008709.1 | - | 2 | 584 | ||
AK044906.1 | - | 2 | 769 | |||
AK049187.1 | - | 2 | 764 | |||
AK132173.1 | - | 2 | 769 | |||
BC049186.1 | - | 2 | 400 | 6485551 | ||
BC058098.1 | - | 2 | 704 | 6824970 | ||
U65079.1 | - | 2 | 621 |