Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epb4.1l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199265.1 | NP_001186194.1 | 6 | 1048 | ||
NM_013511.3 | NP_038539.2 | 6 | 939 | |||
GenBank mRNA | AB370258.1 | - | 5 | 850 | ||
AB370259.1 | - | 5 | 850 | |||
AB370260.1 | - | 5 | 850 | |||
AB370263.1 | - | 5 | 850 | |||
AF044312.1 | - | 5 | 850 | |||
AJ428901.1 | - | 3 | 135 | |||
AJ539142.1 | - | 5 | 850 | |||
AJ539143.1 | - | 5 | 850 | |||
AJ539144.1 | - | 5 | 850 | |||
AJ542537.1 | - | 5 | 850 | |||
AK043148.1 | - | 5 | 827 | |||
AK144088.1 | - | 6 | 1049 | |||
AK167434.1 | - | 6 | 965 | |||
AK170275.1 | - | 6 | 991 | |||
BC023220.1 | - | 6 | 935 | |||
BC023768.1 | - | 6 | 961 | |||
BC026708.1 | - | 6 | 920 | |||
BC026714.1 | - | 5 | 532 | |||
BC029938.1 | - | 6 | 1077 | |||
KF534728.1 | - | 5 | 850 |