Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epb4.1l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013813.1 | NP_038841.1 | 9 | 1121 | ||
GenBank mRNA | AB370264.1 | - | 8 | 1009 | ||
AB523362.1 | - | 8 | 1009 | |||
AB523363.1 | - | 8 | 1009 | |||
AF152247.1 | - | 9 | 1121 | |||
AF177146.1 | - | 8 | 996 | |||
AK031156.1 | - | 9 | 1153 | |||
AK053532.1 | - | 9 | 1122 | |||
AK173080.1 | - | 4 | 408 | |||
BC079858.1 | - | 9 | 1185 | |||
BC152548.1 | - | 9 | 1086 | 40109749 | ||
HG738869.1 | - | 2 | 120 | |||
HG738870.1 | - | 8 | 1009 |