Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epb4.1l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013813.1 | NP_038841.1 | 15 | 1827 | ||
GenBank mRNA | AB370264.1 | - | 14 | 1715 | ||
AB523362.1 | - | 14 | 1661 | |||
AF152247.1 | - | 15 | 1827 | |||
AF177146.1 | - | 13 | 1612 | |||
AK173080.1 | - | 11 | 1588 | |||
BC079858.1 | - | 14 | 1855 | |||
BC152548.1 | - | 14 | 1756 | 40109749 | ||
HG738869.1 | - | 7 | 790 | |||
HG738870.1 | - | 13 | 1679 |